Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synj2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022599.2 | NP_072121.2 | 1 - 2 | 147 | 86 | |
GenBank mRNA | AF260258.1 | - | 1 - 2 | 147 | 86 | |
BC126067.1 | - | 1 - 2 | 135 | 7323715 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |