Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1galt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022950.1 | NP_075239.1 | 1 - 2 | 133 | 69 | |
GenBank mRNA | AF157963.1 | - | 1 - 2 | 133 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |