Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024360.3 | NP_077336.3 | 2 - 3 | 406 | 105 | |
GenBank mRNA | BC061730.1 | - | 2 - 3 | 406 | 5598665 | 105 |
D13417.1 | - | 2 - 3 | 444 | 105 | ||
L04527.1 | - | 968 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |