Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024388.2 | NP_077364.2 | 1 - 2 | 114 | 108 | |
GenBank mRNA | BC097313.1 | - | 1 - 2 | 107 | 7458540 | 108 |
CB581975.1 | - | 1 - 2 | 114 | 108 | ||
U17254.1 | - | 1 - 2 | 107 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |