Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grpel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024487.4 | NP_077813.1 | 1 - 2 | 76 | 85 | |
GenBank mRNA | BC097312.1 | - | 1 - 2 | 74 | 7387647 | 85 |
FQ220295.1 | - | 1 - 2 | 67 | 85 | ||
U62940.1 | - | 1 - 2 | 77 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |