Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031355.1 | NP_112645.1 | 1 - 2 | 83 | 93 | |
XM_006253358.2 | XP_006253420.1 | 1 - 2 | 83 | 93 | ||
GenBank mRNA | AB039664.1 | - | 1 - 2 | 83 | 93 | |
FQ216638.1 | - | 1 - 2 | 68 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Inventoried Probe spans exons |