Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031553.2 | NP_113741.1 | 2 - 3 | 212 | 87 | |
XM_006241196.2 | XP_006241258.1 | 2 - 3 | 230 | 87 | ||
GenBank mRNA | BC089791.1 | - | 2 - 3 | 212 | 7380395 | 87 |
M55045.1 | - | 1 - 2 | 81 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |