Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031557.2 | NP_113745.1 | 3 - 4 | 600 | 71 | |
GenBank mRNA | BC061814.1 | - | 3 - 4 | 600 | 5598666 | 71 |
U53855.1 | - | 3 - 4 | 531 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |