Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031793.1 | NP_113981.1 | 2 - 3 | 247 | 136 | |
XM_006234324.2 | XP_006234386.1 | 2 - 3 | 623 | 136 | ||
XM_006234325.2 | XP_006234387.1 | 2 - 3 | 444 | 136 | ||
XM_008761930.1 | XP_008760152.1 | 1 - 2 | 543 | 136 | ||
GenBank mRNA | AF043642.1 | - | 2 - 3 | 247 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons |