Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031987.1 | NP_114193.1 | 5 - 6 | 601 | 78 | |
XM_006236004.2 | XP_006236066.1 | 5 - 6 | 761 | 78 | ||
XM_006236005.2 | XP_006236067.1 | 6 - 7 | 2091 | 78 | ||
GenBank mRNA | BC074004.1 | - | 5 - 6 | 621 | 7098081 | 78 |
J02844.1 | - | 1042 | 78 | |||
U26033.1 | - | 5 - 6 | 601 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |