Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052802.1 | NP_434689.1 | 3 - 4 | 465 | 102 | |
GenBank mRNA | U25808.1 | - | 3 - 4 | 465 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Rat Androgen Panel 1700 Inventoried Probe spans exons |