Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053463.1 | NP_445915.1 | 3 - 4 | 377 | 71 | |
XM_006229165.1 | XP_006229227.1 | 4 - 5 | 494 | 71 | ||
GenBank mRNA | AF336828.1 | - | 3 - 4 | 377 | 71 | |
BC100643.1 | - | 4 - 5 | 427 | 7930904 | 71 | |
Z36277.1 | - | 4 - 5 | 439 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |