Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053511.1 | NP_445963.1 | 4 - 5 | 688 | 84 | |
XM_008764298.1 | XP_008762520.1 | 5 - 6 | 891 | 84 | ||
GenBank mRNA | AF098301.1 | - | 4 - 5 | 688 | 84 | |
FQ214021.1 | - | 4 - 5 | 681 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |