Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5ar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053619.1 | NP_446071.1 | 1 - 2 | 44 | 62 | |
GenBank mRNA | AB003042.1 | - | 1 - 2 | 44 | 62 | |
BC078770.1 | - | 1 - 2 | 27 | 7121456 | 62 | |
Y09613.1 | - | 1 - 2 | 28 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Rattus norvegicus Rat Chemokine Panel Inventoried Probe spans exons |