Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053638.1 | NP_446090.1 | 9 - 10 | 919 | 76 | |
GenBank mRNA | AB047541.1 | - | 9 - 10 | 919 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |