Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053968.3 | NP_446420.1 | 2 - 3 | 158 | 58 | |
GenBank mRNA | BC058453.1 | - | 2 - 3 | 161 | 6887014 | 58 |
S65838.1 | - | 2 - 3 | 156 | 58 | ||
X89603.1 | - | 2 - 3 | 95 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Hypoxia Panel 1700 Inventoried Probe spans exons |