Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grifin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057187.2 | NP_476535.1 | 3 - 4 | 261 | 75 | |
XM_008768938.1 | XP_008767160.1 | 3 - 4 | 315 | 75 | ||
GenBank mRNA | AF082160.1 | - | 3 - 4 | 261 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |