Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080777.2 | NP_542955.2 | 1 - 2 | 183 | 78 | |
GenBank mRNA | BC087579.1 | - | 1 - 2 | 183 | 7314241 | 78 |
D17764.1 | - | 1 - 2 | 40 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |