Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130429.1 | NP_569113.1 | 6 - 7 | 719 | 139 | |
XM_006233302.2 | XP_006233364.1 | 6 - 7 | 2038 | 139 | ||
XM_006233303.2 | XP_006233365.1 | 6 - 7 | 2037 | 139 | ||
XM_006233309.2 | XP_006233371.1 | 6 - 7 | 1471 | 139 | ||
XM_008761489.1 | XP_008759711.1 | 6 - 7 | 2040 | 139 | ||
XM_008761491.1 | XP_008759713.1 | 6 - 7 | 734 | 139 | ||
XM_008761492.1 | XP_008759714.1 | 6 - 7 | 1471 | 139 | ||
GenBank mRNA | AF198533.1 | - | 6 - 7 | 719 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Rat Wnt Panel Inventoried Probe spans exons |