Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130812.3 | NP_570825.1 | 1 - 2 | 316 | 112 | |
XM_006238380.2 | XP_006238442.1 | 1 - 2 | 248 | 112 | ||
XM_006238381.1 | XP_006238443.1 | 1 - 2 | 248 | 112 | ||
GenBank mRNA | AF474978.1 | - | 1 - 2 | 230 | 112 | |
AF474979.1 | - | 1 - 2 | 232 | 112 | ||
BC086995.1 | - | 1 - 2 | 251 | 7134283 | 112 | |
CK603883.1 | - | 1 - 2 | 254 | 112 | ||
S79760.1 | - | 1 - 2 | 291 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |