Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130822.1 | NP_570835.1 | 2 - 3 | 63 | ||
XM_008769968.1 | XP_008768190.1 | 2 - 3 | 466 | 63 | ||
XM_008769969.1 | XP_008768191.1 | 2 - 3 | 466 | 63 | ||
XM_008769970.1 | XP_008768192.1 | 2 - 3 | 466 | 63 | ||
XM_008769971.1 | XP_008768193.1 | 2 - 3 | 466 | 63 | ||
XM_008769972.1 | XP_008768194.1 | 2 - 3 | 466 | 63 | ||
XM_008769973.1 | XP_008768195.1 | 2 - 3 | 466 | 63 | ||
XM_008769974.1 | XP_008768196.1 | 2 - 3 | 466 | 63 | ||
XM_008769975.1 | XP_008768197.1 | 2 - 3 | 466 | 63 | ||
XM_008769976.1 | XP_008768198.1 | 2 - 3 | 466 | 63 | ||
XM_008769978.1 | XP_008768200.1 | 2 - 3 | 466 | 63 | ||
XM_008769980.1 | XP_008768202.1 | 2 - 3 | 466 | 63 | ||
XM_008769981.1 | XP_008768203.1 | 2 - 3 | 466 | 63 | ||
XM_008769982.1 | XP_008768204.1 | 2 - 3 | 465 | 63 | ||
XM_008769983.1 | XP_008768205.1 | 2 - 3 | 465 | 63 | ||
XM_008769984.1 | XP_008768206.1 | 2 - 3 | 465 | 63 | ||
GenBank mRNA | AF063103.1 | - | 2 - 3 | 63 | ||
AF081157.1 | - | 2 - 3 | 63 | |||
AF081158.1 | - | 2 - 3 | 63 | |||
AF081159.1 | - | 2 - 3 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Rattus norvegicus 1700 Inventoried Probe spans exons |