Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138825.2 | NP_620180.2 | 1 - 2 | 195 | 66 | |
GenBank mRNA | AF245398.1 | - | 1 - 2 | 195 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |