Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138913.1 | NP_620268.1 | 2 - 3 | 522 | 68 | |
XM_006249382.2 | XP_006249444.1 | 2 - 3 | 566 | 68 | ||
GenBank mRNA | AY221507.1 | - | 2 - 3 | 538 | 68 | |
BC086979.1 | - | 2 - 3 | 509 | 7191865 | 68 | |
FQ231421.1 | - | 2 - 3 | 551 | 68 | ||
FQ231433.1 | - | 2 - 3 | 527 | 68 | ||
FQ232784.1 | - | 2 - 3 | 552 | 68 | ||
Z18877.1 | - | 2 - 3 | 540 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |