Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139187.2 | NP_631926.1 | 1 - 2 | 60 | 86 | |
GenBank mRNA | AF416564.2 | - | 1 - 2 | 60 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Inventoried Probe spans exons |