Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145787.2 | NP_665730.2 | 2 | 503 | 60 | |
GenBank mRNA | AJ311952.1 | - | 1 | 449 | 60 | |
BC089774.1 | - | 2 | 503 | 7321342 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon |