Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172019.2 | NP_742016.2 | 1 - 2 | 149 | 82 | |
GenBank mRNA | AF508023.1 | - | 1 - 2 | 166 | 82 | |
BC066660.1 | - | 1 - 2 | 120 | 5623225 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |