Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172091.2 | NP_742088.1 | 10 - 11 | 1083 | 69 | |
NM_172092.2 | NP_742089.1 | 10 - 11 | 1091 | 69 | ||
NR_073147.1 | 9 - 10 | 984 | 69 | |||
XM_006247876.2 | XP_006247938.1 | 10 - 11 | 1197 | 69 | ||
XM_006247877.1 | XP_006247939.1 | 8 - 9 | 922 | 69 | ||
XM_008768468.1 | XP_008766690.1 | 9 - 10 | 1140 | 69 | ||
XM_008768469.1 | XP_008766691.1 | 9 - 10 | 1143 | 69 | ||
GenBank mRNA | BC081700.1 | - | 9 - 10 | 948 | 7192550 | 69 |
CV117246.1 | - | 6 - 7 | 607 | 69 | ||
L04796.1 | - | 10 - 11 | 1053 | 69 | ||
M96674.1 | - | 10 - 11 | 1093 | 69 | ||
X68692.1 | - | 10 - 11 | 1053 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |