Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172327.2 | NP_758830.2 | 1 - 2 | 103 | 60 | |
XM_006230699.2 | XP_006230761.1 | 2 - 3 | 301 | 60 | ||
XM_006230700.2 | XP_006230762.1 | 2 - 3 | 270 | 60 | ||
GenBank mRNA | AF543560.1 | - | 1 - 2 | 137 | 60 | |
BC089110.1 | - | 1 - 2 | 103 | 7311852 | 60 | |
FQ214270.1 | - | 1 - 2 | 211 | 60 | ||
FQ228146.1 | - | 1 - 2 | 131 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |