Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6pc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176077.3 | NP_788266.2 | 3 - 4 | 589 | 60 | |
GenBank mRNA | AY186240.1 | - | 4 - 5 | 419 | 60 | |
BC078689.1 | - | 3 - 4 | 589 | 7134166 | 60 | |
FQ231610.1 | - | 3 - 4 | 485 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |