Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107717.1 | NP_001101187.1 | 4 - 5 | 903 | 73 | |
XM_006233159.2 | XP_006233221.1 | 5 - 6 | 911 | 73 | ||
XM_006233160.2 | XP_006233222.1 | 5 - 6 | 904 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |