Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178333.1 | NP_848016.1 | 15 - 16 | 1912 | 62 | |
XM_008773069.1 | XP_008771291.1 | 17 - 18 | 3242 | 62 | ||
XM_008773070.1 | XP_008771292.1 | 17 - 18 | 3233 | 62 | ||
GenBank mRNA | AB098162.1 | - | 15 - 16 | 1912 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |