Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cela2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012553.2 | NP_036685.1 | 7 - 8 | 816 | 69 | |
GenBank mRNA | FQ226773.1 | - | 7 - 8 | 818 | 69 | |
FQ228000.1 | - | 7 - 8 | 818 | 69 | ||
FQ228239.1 | - | 6 - 7 | 699 | 69 | ||
V01233.1 | - | 7 - 8 | 816 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease 3' Most Rattus norvegicus 1700 Probe spans exons |