Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012735.2 | NP_036867.1 | 16 - 17 | 2571 | 93 | |
XM_006236711.2 | XP_006236773.1 | 15 - 16 | 3640 | 93 | ||
GenBank mRNA | AF027179.1 | - | 16 - 17 | 2375 | 93 | |
M68971.1 | - | 16 - 17 | 2572 | 93 | ||
M68972.1 | - | 8 - 9 | 1173 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |