Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021663.2 | NP_067695.1 | 10 - 11 | 1118 | 72 | |
XM_006230095.2 | XP_006230157.1 | 9 - 10 | 1026 | 72 | ||
GenBank mRNA | AF238223.1 | - | 8 - 9 | 905 | 72 | |
AF250142.1 | - | 9 - 10 | 1005 | 72 | ||
BC061778.1 | - | 10 - 11 | 1118 | 5621784 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |