Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034198.2 | NP_001029370.1 | 5 - 6 | 560 | 76 | |
NM_021671.3 | NP_067703.1 | 5 - 6 | 560 | 76 | ||
GenBank mRNA | AB006135.1 | - | 5 - 6 | 518 | 76 | |
CK359233.1 | - | 5 - 6 | 441 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |