Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022855.2 | NP_074046.1 | 3 | 1050 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Rattus norvegicus Amplicon spans exons and probe does not span exons |