Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crisp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022859.1 | NP_074050.1 | 3 - 4 | 186 | 98 | |
GenBank mRNA | M31173.1 | - | 3 - 4 | 186 | 98 | |
X04643.1 | - | 3 - 4 | 158 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons |