Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Musk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031061.1 | NP_112323.1 | 9 - 10 | 1302 | 95 | |
GenBank mRNA | U34985.1 | - | 9 - 10 | 1302 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Rattus norvegicus Probe spans exons |