Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031087.2 | NP_112349.2 | 8 - 9 | 1070 | 63 | |
XM_006250405.2 | XP_006250467.1 | 10 - 11 | 1221 | 63 | ||
XM_006250407.2 | XP_006250469.1 | 9 - 10 | 1131 | 63 | ||
XM_006250408.2 | XP_006250470.1 | 8 - 9 | 1046 | 63 | ||
XM_008769876.1 | XP_008768098.1 | 9 - 10 | 1813 | 63 | ||
XM_008769877.1 | XP_008768099.1 | 10 - 11 | 1361 | 63 | ||
XM_008769878.1 | XP_008768100.1 | 8 - 9 | 2439 | 63 | ||
GenBank mRNA | AB004454.1 | - | 7 - 8 | 980 | 63 | |
BC078805.1 | - | 8 - 9 | 1070 | 7109917 | 63 | |
D83700.1 | - | 7 - 8 | 973 | 63 | ||
FQ219614.1 | - | 10 - 11 | 1191 | 63 | ||
X99267.1 | - | 8 - 9 | 1093 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |