Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031145.1 | NP_112407.1 | 2 - 3 | 127 | 85 | |
GenBank mRNA | AF136585.1 | - | 2 - 3 | 127 | 85 | |
BC091143.1 | - | 2 - 3 | 113 | 7308828 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |