Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031564.1 | NP_113752.1 | 2 - 3 | 192 | 78 | |
XM_006247976.2 | XP_006248038.1 | 2 - 3 | 276 | 78 | ||
XM_006247977.2 | XP_006248039.1 | 2 - 3 | 1681 | 78 | ||
GenBank mRNA | U69278.1 | - | 2 - 3 | 192 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons |