Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031589.2 | NP_113777.2 | 6 - 7 | 1153 | 65 | |
XM_008766151.1 | XP_008764373.1 | 5 - 6 | 914 | 65 | ||
XM_008766152.1 | XP_008764374.1 | 5 - 6 | 979 | 65 | ||
XM_008766153.1 | XP_008764375.1 | 5 - 6 | 914 | 65 | ||
GenBank mRNA | AF080468.1 | - | 5 - 6 | 983 | 65 | |
BC070889.1 | - | 6 - 7 | 1153 | 7098680 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |