Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sv2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031593.1 | NP_113781.1 | 1 - 2 | 121 | 85 | |
GenBank mRNA | AF060174.1 | - | 1 - 2 | 121 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |