Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031594.1 | NP_113782.1 | 10 - 11 | 1147 | 97 | |
GenBank mRNA | BC078792.1 | - | 10 - 11 | 1088 | 7123247 | 97 |
U32497.1 | - | 10 - 11 | 1124 | 97 | ||
U47031.1 | - | 10 - 11 | 1077 | 97 | ||
X87763.1 | - | 10 - 11 | 1147 | 97 | ||
X91200.1 | - | 10 - 11 | 1062 | 97 | ||
X93565.1 | - | 10 - 11 | 1234 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |