Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zranb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031616.1 | NP_113804.1 | 1 - 2 | 77 | 98 | |
GenBank mRNA | AF013965.1 | - | 1 - 2 | 317 | 98 | |
AF013967.1 | - | 1 - 2 | 77 | 98 | ||
BC087012.1 | - | 1 - 2 | 63 | 7189430 | 98 | |
FQ211774.1 | - | 1 - 2 | 103 | 98 | ||
FQ211917.1 | - | 1 - 2 | 103 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |