Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031707.1 | NP_113895.1 | 2 - 3 | 452 | 113 | |
XM_006231775.2 | XP_006231837.1 | 2 - 3 | 1033 | 113 | ||
XM_008760635.1 | XP_008758857.1 | 2 - 3 | 1020 | 113 | ||
GenBank mRNA | AB003726.1 | - | 2 - 3 | 828 | 113 | |
AB007688.1 | - | 2 - 3 | 459 | 113 | ||
AB017140.1 | - | 2 - 3 | 452 | 113 | ||
AF030088.2 | - | 2 - 3 | 113 | |||
AF093267.1 | - | 2 - 3 | 897 | 113 | ||
AF093268.1 | - | 2 - 3 | 889 | 113 | ||
AJ276327.1 | - | 2 - 3 | 172 | 113 | ||
AJ276328.1 | - | 2 - 3 | 172 | 113 | ||
U92079.1 | - | 2 - 3 | 724 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |