Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031779.2 | NP_113967.1 | 3 - 4 | 1647 | 75 | |
XM_006231185.2 | XP_006231247.1 | 2 - 3 | 1780 | 75 | ||
XM_008760312.1 | XP_008758534.1 | 2 - 3 | 1780 | 75 | ||
GenBank mRNA | AF029105.1 | - | 1867 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |