Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031792.1 | NP_113980.1 | 6 - 7 | 713 | 104 | |
XM_008762386.1 | XP_008760608.1 | 6 - 7 | 447 | 104 | ||
GenBank mRNA | AF043345.2 | - | 6 - 7 | 713 | 104 |
Gene Symbol: LOC100911109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749611.3 | XP_003749659.1 | 855 | 104 | ||
XM_008762411.1 | XP_008760633.1 | 447 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |