Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031792.1 | NP_113980.1 | 9 - 10 | 1019 | 62 | |
XM_008762386.1 | XP_008760608.1 | 9 - 10 | 753 | 62 | ||
GenBank mRNA | AF043345.2 | - | 9 - 10 | 1019 | 62 |
Gene Symbol: LOC100911109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749611.3 | XP_003749659.1 | 1161 | 62 | ||
XM_008762411.1 | XP_008760633.1 | 753 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |