Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031808.1 | NP_113996.1 | 8 - 9 | 1311 | 64 | |
GenBank mRNA | AF067793.1 | - | 8 - 9 | 1311 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Rattus norvegicus Probe spans exons |